ContactUsLinkComponent
SC 150-S21 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S21, Amb+13 to 150 C, 7-17 L VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 30 0 MBAR USB 1 * 1 items
Catalog # HAKA1531211
SC 150-S21 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S21, Amb+13 to 150 C, 7-17 L VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 30 0 MBAR USB 1 * 1 items
Catalog # HAKA1531211
Frequently Bought Together