ContactUsLinkComponent
AC 200 (230V/50Hz) Immersion circulators with adjustable bridge Amb+13 to 200 C, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 M BAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES, Safety class 3 (FL), 1 * 1 items
Catalog # HAKA1560021
AC 200 (230V/50Hz) Immersion circulators with adjustable bridge Amb+13 to 200 C, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 M BAR(330 MBAR SUCTION) REMOTE SENSOR PORT USB RAMP PROGRAM MULTIFUNCTION PORT 8 LANGUAGES, Safety class 3 (FL), 1 * 1 items
Catalog # HAKA1560021
Frequently Bought Together