Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REFRACTOMETER DIGITAL RFM960-T Touch Display NEW (CH PLUG) 1 * 1 items
Catalog # 635-0738
Supplier:
BELLINGHAM STANLEY
Expand image
REFRACTOMETER DIGITAL RFM960-T Touch Display NEW (CH PLUG) 1 * 1 items
Catalog # 635-0738
Supplier:
BELLINGHAM STANLEY
Related Information
Documentation
Videos
21 CFR part 11 Complaince using Polarimeters and Refractometers from Bellingham Sta
Refractometer calibration with B+S calibration fluid
Refractometer zero calibration with distilled water
Touchscreen technology from Bellingham Stanley, a Xylem brand
Datasheet
TDS Polarimeters and Refractometers
Documentation
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals