ContactUsLinkComponent
VACUUM CLEANER MODEL EMI-CWR 120V - INCLUDES ACCESSORIES(S/S WANDS & HOSE ENDS) Technical features: Volt: 120V Hertz: 60 Hz Watt: 1 Kw Ampere: 8.5A Dry recovery Air Flow: 204m /h Vacuum at sealed orifice: 2413 mmH2O Recovery capacity: Dry: with 1 * 1 items
Catalog # TIGV110110A
Supplier:  TIGER-VAC
VACUUM CLEANER MODEL EMI-CWR 120V - INCLUDES ACCESSORIES(S/S WANDS & HOSE ENDS) Technical features: Volt: 120V Hertz: 60 Hz Watt: 1 Kw Ampere: 8.5A Dry recovery Air Flow: 204m /h Vacuum at sealed orifice: 2413 mmH2O Recovery capacity: Dry: with 1 * 1 items
Catalog # TIGV110110A
Supplier:  TIGER-VAC
Frequently Bought Together