ContactUsLinkComponent
pH combination electrode, liquid-filled, pH 0…12, 0 … 80°C 1x ceramic diaphragm, Glass body, S7 Top connector 1 * 1 items
Catalog # HACHLZW5202970002
Supplier:  Hach
pH combination electrode, liquid-filled, pH 0…12, 0 … 80°C 1x ceramic diaphragm, Glass body, S7 Top connector 1 * 1 items
Catalog # HACHLZW5202970002
Supplier:  Hach
Frequently Bought Together