Orders LinkContactUsLinkComponent
Inverted microscope, CKX53
Reflected fluorescence illuminator
Catalog # OLYMN5228400
Supplier:   OLYMPUS OPTICAL
Inverted microscope, CKX53
Catalog # OLYMN5228400
Supplier:  OLYMPUS OPTICAL
Inverted microscope, CKX53
Inverted microscope, CKX53
Reflected fluorescence illuminator
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Ergonomically designed and enhanced imaging for cell culture, the CKX53 is designed for cell sampling and manipulation, especially for fluorescence observations.
  • Observations in lightfield transmission and phase contrast evolutionary to fluorescence
  • High contrast thanks to integrated phase system (IPC) (without ring change)
  • Three-position fluorescence illuminator: Two sets of filters for excitation (blue and green), empty position, manual shutter and neutral filter slide 6%
Compatible with CKX3-SLP and IX2-SL condenser slides, CKX3-SPLIC, CKX3-SLPAS, IX2-SPLC, IX2-SLPH1, IX2-SLPH2 optical elements, CKX3-MVR object guide, CKX3-HO35DM Petri dish rack, reflective fluorescence illuminator and mercury, metal halide or LED light sources.
 : Included CACHN 10× / 0,25 Ph / IPC lens, power cable, platinum insert, Allen key, power supply, two camera cable clips with screws - contrast-inverted 3-D views, optional.
Specifications
  • Description:
    Reflected fluorescence illuminator
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Dust Cover for CKX Microscopes
    Dust Cover for CKX Microscopes
    OLYMN1153000
    Log in for availability
    1 * 1 items - 38,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Power cable with elbow
    Power cable with elbow
    630-1900
    Log in for availability
    1 * 1 items - 17,00 €
Product Information
  • Catalog #
    Availability
    Price
  • CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53
    CKX3-SLP phase contrast slider, pre-centered for CKX53 and IX53
    OLYMN5228500
    Log in for availability
    1 * 1 items - 114,00 €