Cleanroom markers, Sharpie®
Marker, double bag, Red, +
Supplier: MICRONOVA
Jump to:
Product Details & Documents
The industry standard, original pen style, fine point, permanent marker.
- Marks permanently on most surfaces
- Fade and water resistant ink
- Quick drying, non toxic alcohol-based ink formula
Sharpie® fine point markers write on cold or wet surfaces, plastic bags or disposable labware. Marks will not smear on reusable glass, metal, or porcelain labware but can be scrubbed off. Sterile items are gamma irradiated and suitable for Aseptic ISO 5 and other clean production areas.
Packaging: Double bagged, gamma irradiated.
Documents
Specifications
- Sterile:+
- Colour:Red
- Description:Marker, double bag
- Seq:08