Filter paper for special applications
389 F, 110 mm
BINZ146168
: Ahlstrom
These filter papers for suitable for special applications.
Grade 11: Plain, hardened.
Grade 389F: Plain, for determination of fat in foodstuffs.
Grade 480: Plain, phase separation paper, hydrophobic silicone-impregnated paper.
Grade 918: Black filter paper, stained with a sulfur colouring to reveal the particles of bright colour, detection of fluorine or silicon and mycelium in cultivated mushrooms.
Grade 470: Plain, kieselguhr (diatomite) paper for fine turbidity.
Grade 69/K: Plain, activated carbon, used in laboratory: clarification and brightening of dull and dark urines, for polarimetric sugar determination and industry: filtration of galvanic baths and clarification of colored liquids.
- Grade:389 F
- Seq:26
- Size:110 mm