ContactUsLinkComponent
SONOREX SUPER RK 1028, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 500X300X200 MM 28 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 W, HF-OUTPUT: 300 WEFF. 1 * 1 items
Catalog # BAND322-CH
SONOREX SUPER RK 1028, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 500X300X200 MM 28 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 W, HF-OUTPUT: 300 WEFF. 1 * 1 items
Catalog # BAND322-CH
Frequently Bought Together