ContactUsLinkComponent
SONOREX SUPER RK 510 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 300X240X150 MM 9,7 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEA TING 30-80°C (86-176°F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 640 W, HF-OUTPUT: 160 WEFF. 1 * 1 items
Catalog # BAND321-CH
SONOREX SUPER RK 510 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 300X240X150 MM 9,7 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEA TING 30-80°C (86-176°F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 640 W, HF-OUTPUT: 160 WEFF. 1 * 1 items
Catalog # BAND321-CH
Frequently Bought Together