ContactUsLinkComponent
SONOREX SUPER RK 156, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 500X140X100 MM 6 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAIN B ALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 640 W, HF-OUTPUT: 160 WEFF. 1 * 1 items
Catalog # BAND305-GB
SONOREX SUPER RK 156, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 500X140X100 MM 6 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAIN B ALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 640 W, HF-OUTPUT: 160 WEFF. 1 * 1 items
Catalog # BAND305-GB
Frequently Bought Together