ContactUsLinkComponent
SONOREX SUPER RK 514 BH, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 325X300X200 MM 18,7 L, S/S TIMER 1-15 MIN AND CONTINUOUS, H EATING 30-80°C (86-176°F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 860 W, HF-OUTPUT: 215 WEFF. 1 * 1 items
Catalog # BAND263-GB
SONOREX SUPER RK 514 BH, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 325X300X200 MM 18,7 L, S/S TIMER 1-15 MIN AND CONTINUOUS, H EATING 30-80°C (86-176°F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 860 W, HF-OUTPUT: 215 WEFF. 1 * 1 items
Catalog # BAND263-GB
Frequently Bought Together