ContactUsLinkComponent
SONOREX TECHNIK RM 75 UH, ULTRASONIC BATH, 40 KHZ, 230 V 50/60 HZ, ID 580X500X300 MM 90 L, WELDED TANK 2 MM S/S AISI 316 TI TIME R 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 1X1000 WEFF., WITH OVERFLOW GUT 1 * 1 items
Catalog # BAND8220-CH
SONOREX TECHNIK RM 75 UH, ULTRASONIC BATH, 40 KHZ, 230 V 50/60 HZ, ID 580X500X300 MM 90 L, WELDED TANK 2 MM S/S AISI 316 TI TIME R 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 1X1000 WEFF., WITH OVERFLOW GUT 1 * 1 items
Catalog # BAND8220-CH
Frequently Bought Together