GM 2070, HF-GENERATOR, 230 V 50/60 HZ, HF-OUTPUT MAX. 70 WEFF., FOR HD 2070 1 * 1 items
Catalog # BAND517-GB
Supplier: BANDELIN-ELECTRONIC
GM 2070, HF-GENERATOR, 230 V 50/60 HZ, HF-OUTPUT MAX. 70 WEFF., FOR HD 2070 1 * 1 items
Catalog # BAND517-GB
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection