SONOREX SUPER RK 510, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 300X240X150 MM 9,7 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 640 W, HF-OUTPUT: 160 WEFF. 1 * 1 items
Catalog # BAND327-GB
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together