Process documentaion
APH 110 NET500 Segosoft network converter, converts serial data (RS232) to network protocol (TCP/IP), Ø 6×210 mm, G 788 (inc CD)
MIEL9687440
: MIELE
- Description:APH 110 NET500 Segosoft network converter, converts serial data (RS232) to network protocol (TCP/IP), Ø 6×210 mm
- Seq:09
- For:G 788 (inc CD)
- Cat. No.:MIEL9687440
- W×D×H:Ø 6×210 mm