ContactUsLinkComponent
Rabbit anti-T7 is a primary antibody which binds to T7 and is directly conjugated to ALP, Alkaline Phosphatase. This is of advantage to shorten assay time by no need to use a secondary antibody to T7. 1 * 100 µG
Catalog # AGRIAS152976
Supplier:  Agrisera
Rabbit anti-T7 is a primary antibody which binds to T7 and is directly conjugated to ALP, Alkaline Phosphatase. This is of advantage to shorten assay time by no need to use a secondary antibody to T7. 1 * 100 µG
Catalog # AGRIAS152976
Supplier:  Agrisera