ContactUsLinkComponent
Rabbit anti-V5 is a primary antibody which binds to V5 and is directly conjugated to SureLight® R-Phycoerythrin (R-PE). This is of a dvantage to shorten assay time by no need to use a secondary antibody to V5. 1 * 100 µG
  AGRIAS152969
 :  Agrisera
Rabbit anti-V5 is a primary antibody which binds to V5 and is directly conjugated to SureLight® R-Phycoerythrin (R-PE). This is of a dvantage to shorten assay time by no need to use a secondary antibody to V5. 1 * 100 µG
  AGRIAS152969
 :  Agrisera
Frequently Bought Together