ContactUsLinkComponent
Rabbit anti-HA is a primary antibody which binds to HA and is directly conjugated to DyLight® 488. This is of advantage to shorten a ssay time by no need to use a secondary antibody to HA. 1 * 100 µG
  AGRIAS152959
 :  Agrisera
Rabbit anti-HA is a primary antibody which binds to HA and is directly conjugated to DyLight® 488. This is of advantage to shorten a ssay time by no need to use a secondary antibody to HA. 1 * 100 µG
  AGRIAS152959
 :  Agrisera
Frequently Bought Together