ContactUsLinkComponent
Rabbit anti-HA is a primary antibody which binds to HA and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantag e to shorten assay time by no need to use a secondary antibody to HA. 1 * 100 µG
Catalog # AGRIAS152958
Supplier:  Agrisera
Rabbit anti-HA is a primary antibody which binds to HA and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantag e to shorten assay time by no need to use a secondary antibody to HA. 1 * 100 µG
Catalog # AGRIAS152958
Supplier:  Agrisera