ContactUsLinkComponent
Mouse anti-c-myc is a primary antibody which binds to c-myc tag which is directly conjugated to DyLight® 650. This is of advantage t o shorten assay time by no need to use a secondary antibody to c-myc. DyLight® is a trademark of Thermo Fisher Scient 1 * 100 µG
Catalog # AGRIAS152942
Supplier:  Agrisera
Mouse anti-c-myc is a primary antibody which binds to c-myc tag which is directly conjugated to DyLight® 650. This is of advantage t o shorten assay time by no need to use a secondary antibody to c-myc. DyLight® is a trademark of Thermo Fisher Scient 1 * 100 µG
Catalog # AGRIAS152942
Supplier:  Agrisera