ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine, horseradish peroxidase (AS15 TMB-HRP) is a sensitive reagent that is ready to use for the quantitati ve detection of HRP bound to a solid phase or in free solution. 1 * 10 mL
Catalog # AGRIAS15TMB-HRP-10
Supplier:  Agrisera
3,3',5,5'-Tetramethylbenzidine, horseradish peroxidase (AS15 TMB-HRP) is a sensitive reagent that is ready to use for the quantitati ve detection of HRP bound to a solid phase or in free solution. 1 * 10 mL
Catalog # AGRIAS15TMB-HRP-10
Supplier:  Agrisera