Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VWR Precision balance LPG1002-C , capacity: 1000g, readibility: 0,01g, 160 mm pan , graphical display , factory calibration certi ficate 1 * 1 items
Catalog # 611-3387
Supplier:
VWR Collection
Expand image
VWR Precision balance LPG1002-C , capacity: 1000g, readibility: 0,01g, 160 mm pan , graphical display , factory calibration certi ficate 1 * 1 items
Catalog # 611-3387
Supplier:
VWR Collection
Related Information
Documentation
COC_VWR_PRECISION_BALANCES_LG_SERIES_MODELS_WITH_EXTERNAL_CALIBRATION
Manual
Manual VWR Precision Balance with Graphic Display
Documentation
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals