ContactUsLinkComponent
VWR Precision balance LPG2102-C Capacity: 2200g, readibity: 0,01g, 160 mm pan , graphical display , factory calibration certificate 1 * 1 items
Catalog # 611-3385
Supplier:  VWR Collection
undefined
VWR Precision balance LPG2102-C Capacity: 2200g, readibity: 0,01g, 160 mm pan , graphical display , factory calibration certificate 1 * 1 items
Catalog # 611-3385
Supplier:  VWR Collection
Frequently Bought Together