0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
BUFFER PH 9,00 +/-0,01 @25C (BAG IN BOX) 1 * 5 L
REGEBPH28
:
REAGECON
BUFFER PH 9,00 +/-0,01 @25C (BAG IN BOX) 1 * 5 L
REGEBPH28
:
REAGECON
Frequently Bought Together
Accessories for pH/°C meter, handheld
: Testo
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
: EHRENSTORFER
FA2B glycan (G0B), 2-AA labelled 1 * 100 Pmol
: LUDGER
Levetiracetam impurity G CRS 1 * 10 mg
: EDQM
Oxyquinoline Sulfate, Reference Standard
: USP
Potassium heptaiodobismuthate 6,1% in aqueous L(+)-tartaric acid solution with potassium iodide Reag. Ph. Eur. 1070601
: VWR Chemicals
Benzalkonium Chloride (approx. 4% aqueous solution), Reference Standard
: USP
Carfentrazone-ethyl certified 1 * 10 mg
: EHRENSTORFER
Cetyl Alcohol, Reference Standard
: USP
S-LEVETIRACETAM HYDROXYBUTYRAMIDE ANALOG 1 * 10 mg
: Merck