SULFUR COMPOUNDS BY SELECTIVE DETECTION FOR ASTM D5623: MULTI-COMPONENT STANDARD CONTAINING 14 SULFUR SPECIES IN BASE FUEL, 2ML AMPO ULE. S @ 1.0µG/G (FOR EACH COMPONENT). 1 * 1 items
Catalog # PROOG-XMSX14-1.0-2
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together