Rotavapor R-300 B-305, no joint., C, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Cold tra p, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300150C051
Supplier: BUCHI
Frequently Bought Together