Premium Hotplate Stirrer, MSH-20A, analog, ceramic-coated plate 180 x 180 mm, with PWM control system, temp. range up to 380C, speed range 80-1500 rpm, stirring capacity: 20 Liter, heating power: 600 W, dim.: 206 x 307 x 99 mm,weight: 2,8 kg, power 1 * 1 items
Catalog # WITGDH.WMH03120
Supplier: witeg Labortechnik
Frequently Bought Together