Basic fuchsin 88%, pure
Plastic drum, 2,5 kg
Supplier: Thermo Fisher Scientific
Basic fuchsin 88%, pure
Catalog # ACRO419140025
Supplier: Thermo Fisher Scientific
CAS Number: 632-99-5
Jump to:
Product Details & Documents
Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
- CAS Number:632-99-5
- Formula:C₂₀H₂₀ClN₃
- MDL Number:MFCD00012569
- Molecular Weight:337.85 g/mol
- Merck Index:12,05686
- Melting Pt:250 °C (dec.)
- Density:1,22 g/cm³ (20 °C)
- Flash Pt:200 °C
- Storage Temperature:Ambient
- EINECS:211-189-6
Specifications
Specification Test Results
- Appearance:Green Powder
- Infrared spectrum:Conforms
- Loss on drying:≤10 % (105°C)
- Dye content:≥83 % (on dry substance)
- UV Lambda max:549 to 555 nm (in 50:50 IMS:water)
- Absorption Ratio P-15/P+15:1.1 to 1.4
- Specific absorbance A (1%/1cm):≥1800 (at lambda max.)
Specifications
- Pack type:Plastic drum
- Pk:2,5 kg



