Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

3-Chlorophenol 99%

Glass bottle, 1 kg
Catalog # ACRO109810010
Irritant
Environment
3-Chlorophenol 99%
Catalog # ACRO109810010
CAS Number:  108-43-0
3-Chlorophenol 99%
3-Chlorophenol 99%

Glass bottle, 1 kg

CAS Number:  108-43-0
Price
647,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:108-43-0
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002256
  • Molecular Weight:128.56 g/mol
  • Merck Index:13,02173
  • Boiling Pt:214 °C (760 mmHg)
  • Melting Pt:33…35 °C
  • Density:1.218 g/cm³
  • Flash Pt:97 °C
  • Storage Temperature:Ambient
  • EINECS:203-582-6
  • UN:2020
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5650 to 1.5670 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Iron (Fe):
    ≤50 ppm
  • Magnesium (Mg):
    ≤100 ppm
  • Potassium (K):
    ≤50 ppm
  • Sodium (Na):
    ≤100 ppm
  • Silicon dioxide (SiO2):
    ≤1000 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.