2-Chlorocyclohexanone 95% stabilised
Plastic bottle, 5 g
Supplier: Thermo Fisher Scientific
2-Chlorocyclohexanone 95% stabilised
Catalog # ACRO108960050
Supplier: Thermo Fisher Scientific
CAS Number: 822-87-7
Jump to:
Product Details & Documents
- CAS Number:822-87-7
- Formula:C₆H₉ClO
- MDL Number:MFCD00001626
- Molecular Weight:132.59 g/mol
- Boiling Pt:82…83 °C (10 mmHg)
- Melting Pt:23 °C
- Density:1.16 g/cm³
- Flash Pt:82 °C
- Storage Temperature:Refrigerator
- EINECS:212-505-5
- ADR:9,
Documents
ACRO10896
SDS
Specifications
Specification Test Results
- Appearance:Clear brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4850 (20°C, 589 nm)
- GC:≥94 %
- Stabilizer:0.2 % CaCO3/MgO (1:1)
Specifications
- Pack type:Plastic bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



