Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Media for Microbiology
Dehydrated Media for Microbiology
Yeast extract
Print
Yeast extract
Yeast Exract, Plastic bottle, 1 kg
Catalog # ACRO451120010
SDS
Supplier:
Thermo Fisher Scientific
Yeast extract
Catalog # ACRO451120010
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Return to Top
Yeast extract
Yeast Exract, Plastic bottle, 1 kg
Specifications
Jump to:
Specifications
CAS Number:
Price
125,00 €
Log in
for availability
1 * 1 kg
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
ACRO45112
Specifications
Specifications
Pack type:
Plastic bottle
Pk:
1 kg
Description:
Yeast Exract
Specifications