Orders LinkContactUsLinkComponent
Microfiber string mops, M-Zone MegaSorb™
MegaSorb™ mop head, 10 oz., irradiated, White, Microcon blue vinyl
  MICRMZCRMS-10IR
 :   MICRONOVA
Microfiber string mops, M-Zone MegaSorb™
  MICRMZCRMS-10IR
 :  MICRONOVA
Microfiber string mops, M-Zone MegaSorb™
Microfiber string mops, M-Zone MegaSorb™
MegaSorb™ mop head, 10 oz., irradiated, White, Microcon blue vinyl
231,00 €
1 * 12 items
A highly absorbent mop that combines the cleaning efficiency of microfibre with the ease of a looped string mop. The tufted strands are comprised of polyester and nylon filaments, designed to pick up liquids and tiny particles in controlled and critical environments.
  • Highly absorbent and low-linting
  • Polyester and nylon tufted microfibre tubes
  • Picks up tiny particles where cotton fibres just roll over
  • White microfibre option includes an additional scrubbing band
  • Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
The M-Zone™ MegaSorb™ Microfibre String Mop features distinctive microfiber tubes with tufted strands made of polyester and nylon filaments, 30 times smaller than cotton fibres, that work to pick up tiny particles.
  • Colour:
    White
  • Description:
    MegaSorb™ mop head, 10 oz., irradiated
  • Material:
    Microcon blue vinyl
  • Seq:
    02
Accessories
  •  
    Description
    Length
  • MICRMHC-1
    MicroMop® handle, Fibreglass
    1524 mm
    1 * 1 items - 72,20 €
  •  
    Description
  • 129-0170
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    1 * 1 items - 157,00 €
  •  
    Description
    Length
    Material
  • 129-0551
    Stainless steel quick disconnect extendable handle
    965 - 1829 mm
    Stainless steel 304
    1 * 1 items - 358,00 €
  •  
    Description
    Length
  • 129-0165
    MicroMop® handle, Fibreglass/PP
    1626 mm
    1 * 1 items - 85,50 €