Orders LinkContactUsLinkComponent
Counting chambers, Cellometer®
Counting chambers, Cellometer®
Counting chambers with protective film
  734-2886
Counting chambers, Cellometer®
  734-2886
Counting chambers, Cellometer®
Counting chambers, Cellometer®
Counting chambers with protective film
898,00 €
1 * 75 items
These counting chambers can be inserted into a cell counter, so that the cells are imaged directly.
  • Two independent enclosed chambers
  • Compatible with all Cellometer® cell counters
  • Cell suspension (20 μl) loaded via a a single channel pipette
  • Description:
    Counting chambers with protective film
  • Seq:
    01