Laboratory coats, C851
White, 3XL, 100 cm
Catalog # 113-8366
Supplier: Portwest
Jump to:
Product Details & Documents
V-necked coats made of durable Regal fabric (100% pre-shrunk cotton 3111).
- One chest pocket, two front patch pockets
- Concealed stud front closure
- Back vent
- Durable and comfortable
Weight: 305 g/m²
Specifications
- Clothing Size:3XL
- Colour:White
- Length:100 cm
- Seq:07
- Cat. No.:113-8366
- Sleeve or leg length:Long
- Model:C851
- Garment application:General Purpose|Medical|Laboratory|Clinical
- Gender:Unisex
- Closure type:Press studs / snaps
- Fabric:100% cotton
- Weight:305 g/m²