ContactUsLinkComponent
Laboratory centrifuge LMC-3000, WITHOUT rotors - Speed regulation range: 100–3,000 rpm for centrifuge tubes (1,700 × g) - Speed regulation range 100–2,000 rpm for microtitre plates (560 × g) - Setting resolution: 100 rpm - Rotor imbalance diagnosti w 1 * 1 items
Catalog # BSANBS-010208-AAA
Supplier:  BIOSAN
Laboratory centrifuge LMC-3000, WITHOUT rotors - Speed regulation range: 100–3,000 rpm for centrifuge tubes (1,700 × g) - Speed regulation range 100–2,000 rpm for microtitre plates (560 × g) - Setting resolution: 100 rpm - Rotor imbalance diagnosti w 1 * 1 items
Catalog # BSANBS-010208-AAA
Supplier:  BIOSAN
Frequently Bought Together