Accessories
Discharge tube, with recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
Catalog # 613-5217
Supplier:   Brand
Accessories
Catalog # 613-5217
Supplier:  Brand
Accessories
Accessories
Discharge tube, with recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
Price
81,00 €
Log in for availability
1 * 1 items
Specifications
  • Description:
    Discharge tube, with recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
  • Seq:
    17