Accessories
Discharge tube for Dispensette® S Trace Analysis, Tantalum valve spring, without recirculation valve, 105 mm length, 105 mm
Catalog # 613-5210
Supplier:   Brand
Accessories
Catalog # 613-5210
Supplier:  Brand
Accessories
Accessories
Discharge tube for Dispensette® S Trace Analysis, Tantalum valve spring, without recirculation valve, 105 mm length, 105 mm
Price
15,10 €
Log in for availability
1 * 1 items
Specifications
  • Description:
    Discharge tube for Dispensette® S Trace Analysis, Tantalum valve spring, without recirculation valve, 105 mm length
  • Length:
    105 mm
  • Seq:
    11