Accessories
Discharge tube, without recirculation valve, shape: fine tip, 105 mm length, for Dispensette S 1,2,5 and 10 ml
Catalog # 613-5200
Supplier:   Brand
Accessories
Catalog # 613-5200
Supplier:  Brand
Accessories
Accessories
Discharge tube, without recirculation valve, shape: fine tip, 105 mm length, for Dispensette S 1,2,5 and 10 ml
Price
14,70 €
Log in for availability
1 * 1 items
Specifications
  • Description:
    Discharge tube, without recirculation valve, shape: fine tip, 105 mm length, for Dispensette S 1,2,5 and 10 ml
  • Seq:
    03