Accessories
Telescoping filling tube, FEP, adjusts to various bottle heights, length 70-140 mm for Dispensette® S/Dispensette® Organic 1,2,5 and 10 ml
Catalog # 613-5194
Supplier:   Brand
Accessories
Catalog # 613-5194
Supplier:  Brand
Accessories
Accessories
Telescoping filling tube, FEP, adjusts to various bottle heights, length 70-140 mm for Dispensette® S/Dispensette® Organic 1,2,5 and 10 ml
Price
12,20 €
Log in for availability
1 * 1 items
Specifications
  • Description:
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 70-140 mm for Dispensette® S/Dispensette® Organic 1,2,5 and 10 ml
  • Seq:
    23