ContactUsLinkComponent
PUT-IN TUBE EXTENSION, OUTER DIAM. 60,3 mm, INNER 57 mm, ANGLED, FOR EXHAUST AIR DUCTING, IF NECESSARY FOR CONNECTION BY HOSE, FOR I Fplus 1 * 1 items
Catalog # MEMMB29719IF160PLS
Supplier:  MEMMERT
PUT-IN TUBE EXTENSION, OUTER DIAM. 60,3 mm, INNER 57 mm, ANGLED, FOR EXHAUST AIR DUCTING, IF NECESSARY FOR CONNECTION BY HOSE, FOR I Fplus 1 * 1 items
Catalog # MEMMB29719IF160PLS
Supplier:  MEMMERT
Frequently Bought Together