ContactUsLinkComponent
MidiKros 20cm² MPES 0,65µm 0,75mm FLL X FLL sterile 1 * 1 items
Catalog # SPECD02-E65U-07-S
MidiKros 20cm² MPES 0,65µm 0,75mm FLL X FLL sterile 1 * 1 items
Catalog # SPECD02-E65U-07-S
Frequently Bought Together