VWR Analytical balance LAG124i-M , Capacity:120g, Readability: 0.1mg, Graphic display, internal calibration, RS232 serial interfac e, pan size dia 80 mm, 3 door draught shield, GPL procedures, formulation, animal weighing, checkweighing, percentual w 1 * 1 items
Catalog # 611-3265
Supplier: VWR Collection
Frequently Bought Together