Square cell 10 Optical path length 10 mm, sample amount 1.2 mL, without cell lid 1 * 1 items
ATAGRE-82103
: Atago
Square cell 10 Optical path length 10 mm, sample amount 1.2 mL, without cell lid 1 * 1 items
ATAGRE-82103
: Atago
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection