Orders LinkContactUsLinkComponent
Buckets, Rectangular
Buckets, Rectangular
Blue, 15 L
Catalog # PPSA3550010
Buckets, Rectangular
Catalog # PPSA3550010
Buckets, Rectangular
Buckets, Rectangular
Blue, 15 L
Price
68,41 €
Log in for availability
1 * 1 items
Product Details & Documents
Standard bucket, rectangular, made of PP, with stainless steel handle, for Clino® system trolleys.
  • With scaling 5 to15 L
  • Coloured to facilitate use as part of a colour coded system
  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable (approximately 15 to 30 cycles)
Documents
Specifications
  • Capacity:
    15 L
  • Colour:
    Blue
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cover for 15 l buckets, transparent
    Cover for 15 l buckets, transparent
    PPSA2000155
    Log in for availability
    1 * 1 items - 44,91 €