ContactUsLinkComponent
SPACER PLATE 10 X 10 CM, 1.5 MM SPACERS 10 x 10 cm glass plate with integrated spacers 1.5 mm for PerfectBlue Twin S horizontal gel systems. 1 * 1 items
Catalog # 700-0426
Supplier:  VWR Collection
SPACER PLATE 10 X 10 CM, 1.5 MM SPACERS 10 x 10 cm glass plate with integrated spacers 1.5 mm for PerfectBlue Twin S horizontal gel systems. 1 * 1 items
Catalog # 700-0426
Supplier:  VWR Collection
Frequently Bought Together