ContactUsLinkComponent
Packed Column Rt-XLSulfur 2m, 2.00mm ID, 1/8 OD, Sulfinert Tubing Varian 3800 Column Does Not Include Ferrules and Nuts, Installatio n Kits Ordered Seperately 1 * 1 items
Catalog # REST80485-821
Supplier:  Restek
Packed Column Rt-XLSulfur 2m, 2.00mm ID, 1/8 OD, Sulfinert Tubing Varian 3800 Column Does Not Include Ferrules and Nuts, Installatio n Kits Ordered Seperately 1 * 1 items
Catalog # REST80485-821
Supplier:  Restek
Frequently Bought Together