Packed Column 3% Rtx-1 Silcoport W 100/120 6ft, 2mm ID, SS, Shimadzu 14A 2014 1 * 1 items
REST80441-850
: Restek
Packed Column 3% Rtx-1 Silcoport W 100/120 6ft, 2mm ID, SS, Shimadzu 14A 2014 1 * 1 items
REST80441-850
: Restek
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL