ContactUsLinkComponent
Packed Column OPN on ResSil C 80/100 12', 2mm ID Silco Agilent GC NOC 1 * 1 items
Catalog # REST80437-810901
Supplier:  Restek
Packed Column OPN on ResSil C 80/100 12', 2mm ID Silco Agilent GC NOC 1 * 1 items
Catalog # REST80437-810901
Supplier:  Restek
Frequently Bought Together