ContactUsLinkComponent
Packed Column Hayesep Q 80/100, 2m, 2mm ID, For Shimadzu 8A 1 * 1 items
Catalog # REST80433-851
Supplier:  Restek
Packed Column Hayesep Q 80/100, 2m, 2mm ID, For Shimadzu 8A 1 * 1 items
Catalog # REST80433-851
Supplier:  Restek
Frequently Bought Together